Fmoc-Asp-OAll ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
N-Fmoc-L-aspartic acid 1-Allyl ester, Fmoc-Asp-allyl ester, N-Fmoc-L-aspartic acid 1-allyl ester, 1-Allyl N-Fmoc-L-aspartate, Fmoc-Asp-OAll, 1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate, N-[(9H-Fluoren-9-ylmethyloxycarbonyl)]-L-aspartic acid 1-Allyl ester
A2894-1G
A2894-5G
TCIAA2894-1GEA
710
SEK
TCIAA2894-1G
TCIAA2894-5G
Fmoc-Asp-OAll ≥98.0% (by HPLC, titration analysis)
Fmoc-Asp-OAll
Formel:
C₂₂H₂₁NO₆ MW: 395,411 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00467715 CAS nummer: 144120-53-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Specific rotation [a]20/D | -22.0 to -26.0 deg (C=1,DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...