Altrenogest ≥98.0%

Leverantör: TCI
Varning

Synonyms: Allyltrenbolone, 17-alpha-Allyl-17-beta-hydroxyestra-4,9,11-trien-3-one

A2892-200MG A2892-1G
TCIAA2892-200MGEA 415 SEK
TCIAA2892-200MG TCIAA2892-1G
Altrenogest ≥98.0%
Altrenogest
Formel: C₂₁H₂₆O₂
MW: 310.44 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00867855
CAS nummer: 850-52-2
EINECS: 212-703-1
Merck Index: 14,00319

Order Now

Specifikation testresultat

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 116.0 to 120.0 deg-C
Specific rotation [a]20/D -72.0 to -77.0 deg (C=0.5, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR