2-Aminooxazole 98,0 GC, T
Leverantör: TCI
Varning
Synonyms: Oxazol-2-amine
A2884-5G A2884-1G
TCIAA2884-5GEA 6250 SEK
TCIAA2884-5G TCIAA2884-1G
2-Aminooxazole 98,0 GC, T
2-Aminooxazole
Formel: C₃H₄N₂O
MW: 84.10 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD07364485
CAS nummer: 4570-45-0
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C