N-Acetyl-DL-phenylalanine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: Ac-DL-Phe-OH

A2864-5G A2864-25G
TCIAA2864-5GEA 215 SEK
TCIAA2864-5G TCIAA2864-25G
N-Acetyl-DL-phenylalanine ≥98.0% (by HPLC, titration analysis)
N-Acetyl-DL-phenylalanine
Formel: C₁₁H₁₃NO₃
MW: 207,229 g/mol
MDL Number: MFCD00063157
CAS nummer: 2901-75-9
EINECS: 220-793-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR