5-Amino-2,4,6-triiodoisophthaloyl Dichloride ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Amino-2,4,6-triiodoisophthaloyl dichloride
A2854-5G
A2854-25G
TCIAA2854-5GEA
1200
SEK
TCIAA2854-5G
TCIAA2854-25G
5-Amino-2,4,6-triiodoisophthaloyl Dichloride ≥98.0%
5-Amino-2,4,6-triiodoisophthaloyl Dichloride
Formel:
C₈H₂Cl₂I₃NO₂ MW: 595,72 g/mol Smältpunkt: 231°C |
MDL Number:
MFCD00270836 CAS nummer: 37441-29-5 EINECS: 417-220-1 UN: 3077 ADR: 9,III |
Specifikation testresultat
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...