3-Aminobenzofuran-2-carboxamide ≥98.0%

Leverantör: TCI EUROPE
Varning

A2832-1G A2832-5G
TCIAA2832-1GEA 550 SEK
TCIAA2832-1G TCIAA2832-5G
3-Aminobenzofuran-2-carboxamide ≥98.0%
3-Aminobenzofuran-2-carboxamide
Formel: C₉H₈N₂O₂
MW: 176,18 g/mol
MDL Number: MFCD00457147
CAS nummer: 54802-10-7

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR