Aniline Hydroiodide ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
|
Varning
|
Synonyms:
Anilinium iodide
A2778-1G
A2778-5G
TCIAA2778-1GEA
950
SEK
TCIAA2778-1G
TCIAA2778-5G
Aniline Hydroiodide ≥98.0% (by HPLC, total nitrogen)
Aniline Hydroiodide
Formel:
C₆H₇N·HI MW: 221.04 g/mol |
CAS nummer:
45497-73-2 EINECS: 256-239-8 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...