6-Acetyl-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Acetyl-2H-benzo[b][1,4]oxazin-3(4H)-one

A2755-5G A2755-25G
TCIAA2755-5GEA 445 SEK
TCIAA2755-5G TCIAA2755-25G
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by HPLC, total nitrogen)
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one
Formel: C₁₀H₉NO₃
MW: 191,19 g/mol
MDL Number: MFCD01321313
CAS nummer: 26518-71-8

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR