3-Amino-5-fluoropyridine ≥97.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
5-Fluoropyridin-3-amine
A2753-1G
A2753-5G
TCIAA2753-1GEA
535
SEK
TCIAA2753-1G
TCIAA2753-5G
3-Amino-5-fluoropyridine ≥97.0% (by GC, titration analysis)
3-Amino-5-fluoropyridine
Formel:
C₅H₅FN₂ MW: 112.11 g/mol |
MDL Number:
MFCD00234073 CAS nummer: 210169-05-4 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...