4-Amino-2,6-dimethylpyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,6-Dimethylpyridin-4-amine

A2749-1G A2749-5G
TCIAA2749-1GEA 1275 SEK
TCIAA2749-1G TCIAA2749-5G
4-Amino-2,6-dimethylpyridine ≥98.0%
4-Amino-2,6-dimethylpyridine
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
Kokpunkt: 246°C
MDL Number: MFCD00130078
CAS nummer: 3512-80-9
EINECS: 222-515-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 191.0 to 195.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR