(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Toxopyrimidine, 4-Amino-2-methyl-5-pyrimidinemethanol, 4-Amino-5-hydroxymethyl-2-methylpyrimidine
A2730-200MG
A2730-1G
TCIAA2730-200MGEA
910
SEK
TCIAA2730-200MG
TCIAA2730-1G
(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥98.0% (by GC, titration analysis)
(4-Amino-2-methyl-5-pyrimidinyl)methanol
Formel:
C₆H₉N₃O MW: 139,1552 g/mol |
MDL Number:
MFCD00849239 CAS nummer: 73-67-6 Merck Index: 14,09561 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...