5'-Amino-2'-methylacetanilide ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Acetamido-4-methylaniline, N-(5-Amino-2-methylphenyl)acetamide, N3-Acetyl-4-methyl-1,3-phenylenediamine
A2724-1G
A2724-5G
TCIAA2724-1GEA
1050
SEK
TCIAA2724-1G
TCIAA2724-5G
5'-Amino-2'-methylacetanilide ≥98.0% (by HPLC, titration analysis)
5'-Amino-2'-methylacetanilide
Formel:
C₉H₁₂N₂O MW: 164,21 g/mol |
MDL Number:
MFCD00047859 CAS nummer: 5434-30-0 EINECS: 226-599-0 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 143.0 to 145.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...