3-Aminotetrahydrofuran ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Tetrahydrofuran-3-amine
A2700-250MG
A2700-1G
TCIAA2700-250MGEA
1075
SEK
TCIAA2700-250MG
TCIAA2700-1G
3-Aminotetrahydrofuran ≥97.0%
3-Aminotetrahydrofuran
Formel:
C₄H₉NO MW: 87,12 g/mol Kokpunkt: 73°C Flampunkt: 49°C |
MDL Number:
MFCD07778394 CAS nummer: 88675-24-5 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...