3-Amino-4-bromo-1H-pyrazole ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

A2688-1G A2688-5G
TCIAA2688-1GEA 390 SEK
TCIAA2688-1G TCIAA2688-5G
3-Amino-4-bromo-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Amino-4-bromo-1H-pyrazole
Formel: C₃H₄BrN₃
MW: 161,99 g/mol
MDL Number: MFCD00082728
CAS nummer: 16461-94-2

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR