2-Amino-5-fluorobenzothiazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Fluorobenzo[d]thiazol-2-amine
A2682-1G
A2682-5G
TCIAA2682-1GEA
1100
SEK
TCIAA2682-1G
TCIAA2682-5G
2-Amino-5-fluorobenzothiazole ≥98.0%
2-Amino-5-fluorobenzothiazole
Formel:
C₇H₅FN₂S MW: 168,19 g/mol |
MDL Number:
MFCD05664560 CAS nummer: 20358-07-0 |
Specifikation testresultat
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 175.0 to 180.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...