2-Amino-4-(trifluoromethyl)benzonitrile ≥98.0% (by GC, total nitrogen)

Leverantör: TCI
Fara

Synonyms: 3-Amino-4-cyanobenzotrifluoride

A2648-1G A2648-5G
TCIAA2648-1GEA 565 SEK
TCIAA2648-1G TCIAA2648-5G
2-Amino-4-(trifluoromethyl)benzonitrile ≥98.0% (by GC, total nitrogen)
2-Amino-4-(trifluoromethyl)benzonitrile
Formel: C₈H₅F₃N₂
Kokpunkt: 127°C
MDL Number: MFCD04974124
CAS nummer: 1483-54-1
EINECS: 216-047-7
UN: 3439
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystalline
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR