3-Amino-1-propanesulphonic acid ≥98.0%

Leverantör: TCI
Varning

Synonyms: Tramiprosate, Homotaurine, 3-Aminopropane-1-sulphonic acid, 3APS

A2602-5G A2602-25G
TCIAA2602-5GEA 1125 SEK
TCIAA2602-5G TCIAA2602-25G
3-Amino-1-propanesulphonic acid ≥98.0%
3-Amino-1-propanesulphonic acid
Formel: C₃H₉NO₃S
MDL Number: MFCD00008225
CAS nummer: 3687-18-1
EINECS: 222-977-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR