Acetazolamide ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2-Acetamido-5-sulphonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulphonamide

A2598-25G A2598-100G
TCIAA2598-25GEA 885 SEK
TCIAA2598-25G TCIAA2598-100G
Acetazolamide ≥98.0% (by HPLC, titration analysis)
Acetazolamide
Formel: C₄H₆N₄O₃S₂
MW: 222.24 g/mol
MDL Number: MFCD00003105
CAS nummer: 59-66-5
EINECS: 200-440-5
Merck Index: 14,00053

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR