3-Amino-3-(3-bromophenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Synonyms:
DL-3-Amino-3-(3-Bromophenyl)propionic acid
A2535-1G
A2535-5G
TCIAA2535-1GEA
375
SEK
TCIAA2535-1G
TCIAA2535-5G
3-Amino-3-(3-bromophenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(3-bromophenyl)propionic acid
Formel:
C₉H₁₀BrNO₂ Smältpunkt: 226°C |
MDL Number:
MFCD00995843 CAS nummer: 117391-50-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...