2-Benzylaniline ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Aminodiphenylmethane, 2-Benzylaniline

A2519-5G A2519-25G
TCIAA2519-5GEA 445 SEK
TCIAA2519-5G TCIAA2519-25G
2-Benzylaniline ≥98.0% (by GC, titration analysis)
2-Benzylaniline
Formel: C₁₃H₁₃N
MW: 183,25 g/mol
Kokpunkt: 172°C
MDL Number: MFCD00007750
CAS nummer: 28059-64-5
EINECS: 248-806-3

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 50.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR