Acetophenone-O-benzoyloxime ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
Synonyms:
Acetophenone O-Benzoyloxime
A2502-1G
A2502-5G
TCIAA2502-1GEA
965
SEK
TCIAA2502-1G
TCIAA2502-5G
Acetophenone-O-benzoyloxime ≥98.0% (by HPLC, total nitrogen)
Acetophenone-O-benzoyloxime
Formel:
C₁₅H₁₃NO₂ |
CAS nummer:
26060-56-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...