Aztreonam ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulpho-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid, 2-((((Z)-1-(2-Aminothiazol-4-yl)-2-(((2S,3S)-2-methyl-4-oxo-1-sulphoazetidin-3-yl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid
A2466-5G
A2466-500MG
TCIAA2466-5GEA
2750
SEK
TCIAA2466-5G
TCIAA2466-500MG
Aztreonam ≥98.0%
Aztreonam
Formel:
C₁₃H₁₇N₅O₈S₂ MW: 435,43 g/mol Smältpunkt: 227°C |
MDL Number:
MFCD00072145 CAS nummer: 78110-38-0 EINECS: 278-839-9 Merck Index: 14,00925 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Specific rotation [a]20/D | -26.0 to -32.0 deg (C=0.5、H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...