2-Amino-6-(trifluoromethyl)benzothiazole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 6-(Trifluoromethyl)benzo[d]thiazol-2-amine

A2461-1G A2461-5G
TCIAA2461-1GEA 325 SEK
TCIAA2461-1G TCIAA2461-5G
2-Amino-6-(trifluoromethyl)benzothiazole ≥98.0%
2-Amino-6-(trifluoromethyl)benzothiazole
Formel: C₈H₅F₃N₂S
MW: 218.20 g/mol
MDL Number: MFCD00269597
CAS nummer: 777-12-8

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 115.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR