3-Aminopiperidin-2-one ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3-Amino-2-piperidinone, 3-Amino-2-piperidone, 2-Amino-delta-valerolactam
A2417-1G A2417-5G
TCIAA2417-1GEA 1900 SEK
TCIAA2417-1G TCIAA2417-5G
3-Aminopiperidin-2-one ≥98.0%
3-Aminopiperidin-2-one
Formel: C5H10N2O
MW: 114,1457 g/mol
Storage Temperature: Frys
MDL Number: MFCD06809610
CAS nummer: 1892-22-4
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %