2-Acetoxycinnamic acid ≥95.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-(2-Acetoxyphenyl)acrylic acid, 3-(2-Acetoxyphenyl)-2-propenoic Acid
A2413-1G
A2413-5G
TCIAA2413-1GEA
245
SEK
TCIAA2413-1G
TCIAA2413-5G
2-Acetoxycinnamic acid ≥95.0% (by GC, titration analysis)
2-Acetoxycinnamic acid
Formel:
C₁₁H₁₀O₄ MW: 206,20 g/mol |
MDL Number:
MFCD00014375 CAS nummer: 55620-18-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 154.0 to 158.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...