N-(6-Aminohexyl)-5-chloro-1-naphthalenesulphonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
W-7 Hydrochloride, N-(6-Aminohexyl)-5-chloronaphthalene-1-sulphonamide hydrochloride
A2409-1G
A2409-100MG
TCIAA2409-1GEA
6550
SEK
TCIAA2409-1G
TCIAA2409-100MG
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulphonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulphonamide hydrochloride
Formel:
C₁₆H₂₁ClN₂O₂S MW: 377,32 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00012559 CAS nummer: 61714-27-0 |
Specifikation testresultat
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...