2-(2-Aminoethyl)-1-methylpyrrolidine ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-(1-Methyl-2-pyrrolidinyl)ethylamine
A2382-1G
A2382-5G
TCIAA2382-1GEA
630
SEK
TCIAA2382-1G
TCIAA2382-5G
2-(2-Aminoethyl)-1-methylpyrrolidine ≥97.0%
2-(2-Aminoethyl)-1-methylpyrrolidine
Formel:
C₇H₁₆N₂ MW: 128,22 g/mol Flampunkt: 65°C |
MDL Number:
MFCD00003175 CAS nummer: 51387-90-7 EINECS: 257-169-0 UN: 2735 ADR: 8,III |
Specifikation testresultat
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...