2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside ≥92.0%

Leverantör: TCI

Synonyms: 2-Azidoethyl 2,3,4,6-Tetra-O-acetyl-β-D-glucopyranoside

A2377-1G A2377-5G
TCIAA2377-1GEA 2375 SEK
TCIAA2377-1G TCIAA2377-5G
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside ≥92.0%
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranoside
Formel: C₁₆H₂₃N₃O₁₀
Storage Temperature: Kylskåp
CAS nummer: 140428-81-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 92.0 area%
Melting point 113.0 to 117.0 deg-C
Specific rotation [a]20/D -33.0 to -40.0 deg (C=1.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR