N-(4-Aminobenzoyl)-β-alanine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 3-(4-Aminobenzamido)propionic Acid, N-(4-Aminobenzoyl)-beta-alanine

A2347-25G A2347-5G
TCIAA2347-25GEA 605 SEK
TCIAA2347-25G TCIAA2347-5G
N-(4-Aminobenzoyl)-β-alanine ≥98.0% (by HPLC, titration analysis)
N-(4-Aminobenzoyl)-beta-alanine
Formel: C₁₀H₁₂N₂O₃
MW: 208,22 g/mol
MDL Number: MFCD00009805
CAS nummer: 7377-08-4

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR