(1S,2S)-(+)-1-Amino-2-indanol ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
(1S,2S)-1-Amino-2-indanol, (1S,2S)-(+)-1-Amino-2-hydroxyindan
A2307-1G
A2307-5G
TCIAA2307-1GEA
655
SEK
TCIAA2307-1G
TCIAA2307-5G
(1S,2S)-(+)-1-Amino-2-indanol ≥98.0%
(1S,2S)-(+)-1-Amino-2-indanol
Formel:
C₉H₁₁NO MW: 149.19 g/mol |
MDL Number:
MFCD08275383 CAS nummer: 163061-74-3 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...