5-Acetylindan ≥98.0%
Leverantör: TCI
A2289-25G
A2289-5G
TCIAA2289-25GEA
4175
SEK
TCIAA2289-25G
TCIAA2289-5G
5-Acetylindan ≥98.0%
5-Acetylindan
Formel:
C₁₁H₁₂O Kokpunkt: 85°C |
MDL Number:
MFCD00021246 CAS nummer: 4228-10-8 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...