N-Acetyl-4-hydroxy-L-proline (cis and trans mixture) cis- and trans-mixture ≥97.0%

Leverantör: TCI
Fara

Synonyms: N-Acetyl-4-hydroxypyrrolidine-2-carboxylic Acid, Ac-Hyp-OH

A2265-5G A2265-25G
TCIAA2265-5GEA 365 SEK
TCIAA2265-5G TCIAA2265-25G
N-Acetyl-4-hydroxy-L-proline (cis and trans mixture) cis- and trans-mixture ≥97.0%
N-Acetyl-4-hydroxy-L-proline (cis and trans mixture)
Formel: C₇H₁₁NO₄
CAS nummer: 926905-71-7
EINECS: 251-780-6

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D -114 to -120 deg (C=4, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR