5-Aza-2'-deoxycytidine
Leverantör: TCI EUROPE
Varning
Synonyms: Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one
A2232-20MG A2232-100MG
TCIAA2232-20MGEA 735 SEK
TCIAA2232-20MG TCIAA2232-100MG
5-Aza-2'-deoxycytidine
5-Aza-2'-deoxycytidine
Formel: C₈H₁₂N₄O₄
MW: 228,21 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00043011
CAS nummer: 2353-33-5
EINECS: 219-089-4
Merck Index: 14,02853
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Elemental analysis (Nitrogen) 23.8 to 25.0 %