2-Amino-4-chloropyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine

A2152-5G A2152-25G
TCIAA2152-5GEA 685 SEK
TCIAA2152-5G TCIAA2152-25G
2-Amino-4-chloropyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-chloropyridine
Formel: C₅H₅ClN₂
MW: 128,56 g/mol
MDL Number: MFCD04113820
CAS nummer: 19798-80-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e1e6d41d1c47ab51714040aed686ce0668df743&vid=1e1e6d41d1c47ab51714040aed686ce0668df743&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR