Artemisinin ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Qinghaosu, Arteannuin, (3R,5aS,6R,8aS,9R,12S,12aR)-3,6,9-Trimethyloctahydro-12H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10(3H)-one

A2118-5G A2118-1G
TCIAA2118-5GEA 1750 SEK
TCIAA2118-5G TCIAA2118-1G
Artemisinin ≥97.0%
Artemisinin
Formel: C₁₅H₂₂O₅
MW: 282,34 g/mol
MDL Number: MFCD00081057
CAS nummer: 63968-64-9
Merck Index: 14,00817
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=296fb654b7796e1f798152654ac2ae72916079d5&vid=296fb654b7796e1f798152654ac2ae72916079d5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 151.0 to 155.0 deg-C
Specific rotation [a]20/D +70.0 to +80.0 deg (C=0.5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR