Amoxicillin trihydrate ≥98.0%

Leverantör: TCI

A2099-25G A2099-5G
TCIAA2099-25GEA 1200 SEK
TCIAA2099-25G TCIAA2099-5G
Amoxicillin trihydrate ≥98.0%
Amoxicillin trihydrate
Formel: C₁₆H₁₉N₃O₅S
MDL Number: MFCD00072029
CAS nummer: 61336-70-7
Merck Index: 14,00577

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +290 to +315 deg (C=0.1, H2O) (calcd.on anh.substance)
Water 11.0 to 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR