N4-Acetylcytosine ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
N-(2-Oxo-1,2-dihydropyrimidin-4-yl)acetamide
A2089-25G
A2089-5G
TCIAA2089-25GEA
830
SEK
TCIAA2089-25G
TCIAA2089-5G
N4-Acetylcytosine ≥98.0%
N4-Acetylcytosine
Formel:
C₆H₇N₃O₂ MW: 153.14 g/mol Smältpunkt: 325°C |
MDL Number:
MFCD00134466 CAS nummer: 14631-20-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...