2-Amino-6-chloropurine riboside ≥95.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
6-Chloroguanineriboside, (2R,3R,4S,5R)-2-(2-amino-6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2-Amino-6-chloro-9-(beta-D-ribofuranosyl)purine
A2054-1G
A2054-5G
TCIAA2054-1GEA
455
SEK
TCIAA2054-1G
TCIAA2054-5G
2-Amino-6-chloropurine riboside ≥95.0% (by HPLC, titration analysis)
2-Amino-6-chloropurine riboside
Formel:
C₁₀H₁₂ClN₅O₄ MW: 301,69 g/mol Smältpunkt: 175°C |
MDL Number:
MFCD00005735 CAS nummer: 2004-07-1 EINECS: 217-905-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Specific rotation [a]20/D | -35.0 to -40.0 deg (C=0.1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...