3-Aminopyridazine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Pyridazin-3-amine
A2048-1G
A2048-5G
TCIAA2048-1GEA
765
SEK
TCIAA2048-1G
TCIAA2048-5G
3-Aminopyridazine ≥98.0% (by GC, titration analysis)
3-Aminopyridazine
Formel:
C4H5N3 MW: 95,10 g/mol |
MDL Number:
MFCD01529869 CAS nummer: 5469-70-5 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 168.0 to 173.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...