2-Acetyl-6-methoxynaphthalene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6'-Methoxy-2'-acetonaphthone

A2030-25G A2030-500G A2030-100G
TCIAA2030-25GEA 310 SEK
TCIAA2030-25G TCIAA2030-500G TCIAA2030-100G
2-Acetyl-6-methoxynaphthalene ≥98.0%
2-Acetyl-6-methoxynaphthalene
Formel: C₁₃H₁₂O₂
MW: 200,24 g/mol
Kokpunkt: 195°C
MDL Number: MFCD00021643
CAS nummer: 3900-45-6
EINECS: 223-453-8

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR