2-Acetamidopyridine ≥98.0%
Leverantör: TCI
A1911-1G
A1911-5G
TCIAA1911-1GEA
575
SEK
TCIAA1911-1G
TCIAA1911-5G
2-Acetamidopyridine ≥98.0%
2-Acetamidopyridine
Formel:
C₇H₈N₂O MW: 136.15 g/mol Kokpunkt: 148°C |
MDL Number:
MFCD00023423 CAS nummer: 5231-96-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...