4-Acetamidopyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: N-(Pyridin-4-yl)acetamide, N-(4-Pyridyl)acetamide

A1883-25G A1883-5G
TCIAA1883-25GEA 2650 SEK
TCIAA1883-25G TCIAA1883-5G
4-Acetamidopyridine ≥98.0% (by GC, titration analysis)
4-Acetamidopyridine
Formel: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD00720126
CAS nummer: 5221-42-1
EINECS: 226-018-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR