4-Methoxybutyl acetate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Acetic Acid 4-Methoxybutyl Ester
A1676-1G A1676-5G
TCIAA1676-1GEA 445 SEK
TCIAA1676-1G TCIAA1676-5G
4-Methoxybutyl acetate ≥98.0%
4-Methoxybutyl acetate
Formel: C₇H₁₄O₃
MW: 146,19 g/mol
Kokpunkt: 71°C
MDL Number: MFCD06797048
CAS nummer: 15057-11-1
UN: 3272
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %