2-Methoxybutyl acetate ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Acetic Acid 2-Methoxybutyl Ester

A1675-25G A1675-5G
TCIAA1675-25GEA 5650 SEK
TCIAA1675-25G TCIAA1675-5G
2-Methoxybutyl acetate ≥97.0%
2-Methoxybutyl acetate
Formel: C₇H₁₄O₃
MW: 146,19 g/mol
MDL Number: MFCD06797047
CAS nummer: 1173168-18-7

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR