4-(4-Aminophenyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Thiomorpholinoaniline 1',1'-Dioxide, 4-(4-Aminophenyl)thiomorpholine 1,1-dioxide, 4-(1,1-Dioxothiomorpholino)aniline

A1669-1G A1669-5G
TCIAA1669-1GEA 1125 SEK
TCIAA1669-1G TCIAA1669-5G
4-(4-Aminophenyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
4-(4-Aminophenyl)thiomorpholine-1,1-dioxide
Formel: C₁₀H₁₄N₂O₂S
MW: 226,29 g/mol
MDL Number: MFCD06797046
CAS nummer: 105297-10-7

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 197.0 deg-C
Solubility in hot Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR