4-Acetamido-1-benzylpiperidine ≥98.0% (by GC, titration analysis)

Leverantör: TCI

Synonyms: N-(1-Benzylpiperidin-4-yl)acetamide

A1654-25G A1654-5G
TCIAA1654-25GEA 2800 SEK
TCIAA1654-25G TCIAA1654-5G
4-Acetamido-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
4-Acetamido-1-benzylpiperidine
Formel: C₁₄H₂₀N₂O
MW: 232.33 g/mol
Smältpunkt: 140°C
MDL Number: MFCD00247343
CAS nummer: 50534-23-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR