4-Acetamidopiperidine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
N-(Piperidin-4-yl)acetamide
A1653-25G
A1653-5G
TCIAA1653-25GEA
4925
SEK
TCIAA1653-25G
TCIAA1653-5G
4-Acetamidopiperidine ≥98.0% (by GC, titration analysis)
4-Acetamidopiperidine
Formel:
C₇H₁₄N₂O MW: 142.20 g/mol Smältpunkt: 141°C |
MDL Number:
MFCD05254805 CAS nummer: 5810-56-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...