Tributyrin, liquid
Leverantör: MP Biomedicals
Synonyms: Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate
0210311191 0210311180 0210311183 0210311190
ICNA0210311191EA 5600 SEK
ICNA0210311191 ICNA0210311180 ICNA0210311183 ICNA0210311190
Tributyrin, liquid
Tributyrin
Tributyrin, is a glycerol substituted with three butyrl esters.
Tributyrin induces differentiation of malignant cells followed by arrest of cell growth and apoptosis. It is used as a deliverable source of natural butyrate and is demonstrated to inhibit growth of and induce cell differentiation of Caco-2 cells more potently than natural butyrate.
Tributyrin induces differentiation of malignant cells followed by arrest of cell growth and apoptosis. It is used as a deliverable source of natural butyrate and is demonstrated to inhibit growth of and induce cell differentiation of Caco-2 cells more potently than natural butyrate.
Soluble in alcohol and ether; insoluble in water.
Formel: C₁₅H₂₆O₆
MW: 302,37 g/mol
Kokpunkt: 287…288 °C (lit.)
Smältpunkt: −75 °C (lit.)
Densitet: 1,035 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00009392
CAS nummer: 60-01-5
EINECS: 200-451-5
Merck Index: 12,09750