2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-Acetamido-2-deoxy-β-D-glucopyranosyl azide 3,4,6-triacetate

A1616-1G A1616-5G
TCIAA1616-1GEA 935 SEK
TCIAA1616-1G TCIAA1616-5G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98.0%
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl azide
Formel: C₁₄H₂₀N₄O₈
MW: 372,33 g/mol
Smältpunkt: 167°C
Storage Temperature: Frys
MDL Number: MFCD00216968
CAS nummer: 6205-69-2

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -43.0 to -48.0 deg (C=1,chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR