Aminoferrocene ≥96.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Ferrocenylamine

A1601-100MG A1601-1G
TCIAA1601-100MGEA 895 SEK
TCIAA1601-100MG TCIAA1601-1G
Aminoferrocene ≥96.0% (by GC, titration analysis)
Aminoferrocene
Formel: C₁₀H₂FeN
MW: 191,97 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD03844760
CAS nummer: 1273-82-1

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 96.0 %
Purity (Chelometric Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR